Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CTRP1/C1qTNF1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10445SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10445SP 25 μg
MAB10445100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10445SP Supplier R&D Systems Supplier No. MAB10445SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

C1qTNF1/CTRP1 Monoclonal specifically detects C1qTNF1/CTRP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen C1qTNF1/CTRP1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 395520
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 1-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9BXJ1
Gene Alias C1q and tumor necrosis factor related protein 1, C1qTNF1, CTRP1G protein-coupled receptor-interacting protein, FLJ90694, G protein coupled receptor interacting protein, GIPcomplement C1q tumor necrosis factor-related protein 1, ZSIG37
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived human CTRP1/C1qTNF1, Arg26-Pro281, Accession # Q9BXJ1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114897
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.