Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CTRP3/C1qTNF3/CORS26 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $409.50
Specifications
Antigen | C1qTNF3/CORS26/CTRP3 |
---|---|
Clone | 860115 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB7925SP
![]() |
R&D Systems
MAB7925SP |
25 μg |
Each for $200.50
|
|
|||||
MAB7925100
![]() |
R&D Systems
MAB7925100 |
100 μg |
Each for $409.50
|
|
|||||
Description
C1qTNF3/CORS26/CTRP3 Monoclonal specifically detects C1qTNF3/CORS26/CTRP3 in Human samples. It is validated for ELISA.Specifications
C1qTNF3/CORS26/CTRP3 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
C1ATNF3, C1q and tumor necrosis factor related protein 3, C1qTNF3, Cartducin, cartonectin, collagenous repeat-containing sequence of 26-kDa, complement C1q tumor necrosis factor-related protein 3, complement-c1q tumor necrosis factor-related protein 3, Corcs, Cors, Cors-26, CORS26, CTRP32310005P21Rik, FLJ37576, Secretory protein CORS26 | |
Mouse myeloma cell line NS0-derived human CTRP3/C1qTNF3/CORS26, Gln23-Lys246, Accession # Q9BXJ4-1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
860115 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q9BXJ4-1 | |
114899 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title