Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CTRP3/C1qTNF3/CORS26 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7925
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7925 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7925 Supplier R&D Systems Supplier No. AF7925
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

C1qTNF3/CORS26/CTRP3 Polyclonal specifically detects C1qTNF3/CORS26/CTRP3 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen C1qTNF3/CORS26/CTRP3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q9BXJ4
Gene Alias C1ATNF3, C1q and tumor necrosis factor related protein 3, C1qTNF3, Cartducin, cartonectin, collagenous repeat-containing sequence of 26-kDa, complement C1q tumor necrosis factor-related protein 3, complement-c1q tumor necrosis factor-related protein 3, Corcs, Cors, Cors-26, CORS26, CTRP32310005P21Rik, FLJ37576, Secretory protein CORS26
Gene Symbols C1QTNF3
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human CTRP3/C1qTNF3/CORS26 Asp24-Lys246 Accession # Q9BXJ4
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114899
Test Specificity Detects human CTRP3/C1qTNF3/CORS26 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse CTRP3/C1qTNF3/CORS26 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.