Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CTRP4/C1qTNF4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3456
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3456 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3456 Supplier R&D Systems Supplier No. AF3456
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

C1qTNF4/CTRP4 Polyclonal specifically detects C1qTNF4/CTRP4 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen C1qTNF4/CTRP4
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Gene Accession No. Q9BXJ3
Gene Alias C1q and tumor necrosis factor related protein 4, C1qTNF4, complement C1q tumor necrosis factor-related protein 4, complement-c1q tumor necrosis factor-related protein 4, CTRP4ZACRP4, ZACRP4
Gene Symbols C1QTNF4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CTRP4/C1qTNF4 Leu17-Leu329 Accession # Q9BXJ3
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114900
Test Specificity Detects human CTRP4/C1qTNF4 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) CTRP5 is observed and less than 1% cross-reactivity with rhCTRP1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.