Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CTRP9/C1qTNF9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $410.00
Specifications
Antigen | CTRP9/C1qTNF9 |
---|---|
Clone | 653533 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
CTRP9/C1qTNF9 Monoclonal specifically detects CTRP9/C1qTNF9 in Human samples. It is validated for Western Blot.Specifications
CTRP9/C1qTNF9 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P0C862 | |
338872 | |
Mouse myeloma cell line NS0-derived recombinant human CTRP9/C1qTNF9 Asn16-Pro333 Accession # P0C862 | |
Primary | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Detects human CTRP9/C1qTNF9 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) C1qTNF1, rhCORS26/C1qTNF3, rhC1qTNF4, rhC1qTNF5, rhC1QL2 is observed. |
653533 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AQL1, C1q and tumor necrosis factor related protein 9, C1qTNF9, C1QTNF9AComplement C1q tumor necrosis factor-related protein 9, complement C1q tumor necrosis factor-related protein 9A, CTRP9, MGC48915 | |
C1QTNF9 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title