Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CTRP9/C1qTNF9 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $468.50
Specifications
Antigen | CTRP9/C1qTNF9 |
---|---|
Dilution | Western Blot 2 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CTRP9/C1qTNF9 Polyclonal specifically detects CTRP9/C1qTNF9 in Human samples. It is validated for Western Blot.Specifications
CTRP9/C1qTNF9 | |
Western Blot | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
AQL1, C1q and tumor necrosis factor related protein 9, C1qTNF9, C1QTNF9AComplement C1q tumor necrosis factor-related protein 9, complement C1q tumor necrosis factor-related protein 9A, CTRP9, MGC48915 | |
Mouse myeloma cell line NS0-derived recombinant human CTRP9/C1qTNF9 Asn16-Pro333 Accession # P0C862 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human CTRP9/C1qTNF9 in direct ELISAs. |
Western Blot 2 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
P0C862 | |
338872 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title