Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CX36 Partial ORF (NP_065711.1, 99 a.a. - 197 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7101523
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene is a member of the connexin gene family that is expressed predominantly in mammalian neurons. Connexins associate in groups of 6 and are organized radially around a central pore to form connexons. Each gap junction intercellular channel is formed by the conjunction of 2 connexons. [provided by RefSeq]

Sequence: HQSAKQRERRYSTVFLALDRDPPESIGGPGGTGGGGSGGGKREDKKLQNAIVNGVLQNTENTSKETEPDCLEVKELTPHPSGLRTASKSKLRRQEGISR

Specifications

Accession Number NP_065711.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57369
Molecular Weight (g/mol) 36.63kDa
Name CX36 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen HQSAKQRERRYSTVFLALDRDPPESIGGPGGTGGGGSGGGKREDKKLQNAIVNGVLQNTENTSKETEPDCLEVKELTPHPSGLRTASKSKLRRQEGISR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CX36/GJA9/MGC138315/MGC138319
Common Name GJD2
Gene Symbol GJD2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.