Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CX3CL1/Fractalkine Chemokine Domain Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC365U100UG
This item is not returnable.
View return policy
Description
CX3CL1/Fractalkine Monoclonal specifically detects CX3CL1/Fractalkine in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CX3CL1/Fractalkine | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P78423 | |
CX3CL1 | |
Mouse myeloma cell line NS0-derived recombinant human CX3CL1/Fractalkine Chemokine Domain Gln25-Arg339 (Ser199Asn) (predicted) Accession # P78423 | |
RUO | |
6376 | |
Human | |
IgG1 |
Flow Cytometry | |
51637 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1 | |
Mouse | |
100 μg | |
Primary | |
Detects human CX3CL1/Fractalkine Chemokine Domain in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CX3CL1 or recombinant rat CX3CL1 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction