Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CX3CL1/Fractalkine Chemokine Domain Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 19 publications
Supplier: R&D Systems AF365
This item is not returnable.
View return policy
Description
CX3CL1/Fractalkine Polyclonal specifically detects CX3CL1/Fractalkine in Human samples. It is validated for Western Blot, Neutralization.Specifications
CX3CL1/Fractalkine | |
Polyclonal | |
Western Blot 0.1 ug/mL, Neutralization 0.3-1.5 ug/mL | |
ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1 | |
Goat | |
Affinity Purified | |
RUO | |
6376 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization | |
Unconjugated | |
Q6I9S9 | |
CX3CL1 | |
E. coli-derived recombinant human CX3CL1/Fractalkine Gln25-Gly100 Accession # Q6I9S9 | |
100 μg | |
Primary | |
Detects human CX3CL1/Fractalkine in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) MCP-2 and rhMCP-4 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction