Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL1/2/3/GRO Pan Specific Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC2761G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC2761G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC2761G100 Supplier R&D Systems Supplier No. IC2761G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL1/2/3/GRO Monoclonal specifically detects CXCL1/2/3/GRO in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CXCL1/2/3/GRO
Applications Flow Cytometry
Classification Monoclonal
Clone 29702
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P19876
Gene Alias CINC1, CINC-1, CINC-2, CINC3, CINC-3, Dcip1, FSP, Gm1960, GRO alpha, GRO beta, GRO gamma, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, MGSA-beta, MIP-2, NAP-3, SCYB1
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL3 Ala35-Asn107 Accession # P19876
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2919
Test Specificity Detects human CXCL1/2/3 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) CXCL2 is observed and no cross-reactivity with recombinant human (rh) VIC, rmVIC, rhCCL3L1, rhCCL4L1, or recombinant canine IL-8 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.