Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL1/2/3/GRO Pan Specific Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2761SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2761SP 25 μg
MAB2761 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2761SP Supplier R&D Systems Supplier No. MAB2761SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL1/2/3/GRO Monoclonal specifically detects CXCL1/2/3/GRO in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen CXCL1/2/3/GRO
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 29702
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P19876
Gene Alias CINC1, CINC-1, CINC-2, CINC3, CINC-3, Dcip1, FSP, Gm1960, GRO alpha, GRO beta, GRO gamma, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, MGSA-beta, MIP-2, NAP-3, SCYB1
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL3 Ala35-Asn107 Accession # P19876
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2919
Test Specificity Detects human CXCL1/2/3 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) CXCL2 is observed and no cross-reactivity with recombinant human (rh) VIC, rmVIC, rhCCL3L1, rhCCL4L1, or recombinant canine IL-8 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.