Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CXCL1/GRO alpha (aa 39-107) Protein

Catalog No. p-7121958
Click to view available options
Formulation:
With BSA as carrier
Without BSA as carrier
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human CXCL1/GRO alpha (aa 39-107) Protein is derived from E. coli. The Recombinant Human CXCL1/GRO alpha (aa 39-107) Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature.

The product is shipped at ambient temperature.

Specifications

Accession Number Q5M8T4
For Use With (Application) Bioactivity
Formulation Without BSA as carrier
Gene ID (Entrez) 1490
Molecular Weight (g/mol) 6 kDa, reducing conditions
Quantity 25 μg
Source E. coli-derived human CXCL1/GRO alpha/KC/CINC-1 protein Thr39-Asn107
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of protein by LAL method
Gene Alias CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1
Biological Activity 0.15 to 0.9ng/mL (ED 50)
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag No
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.