Learn More
Description
Includes
Human CXCL1/GRO-alpha Capture Antibody; Human CXCL1/GRO-alpha Detection Antibody; Human CXCL1/GRO-alpha Standard; Streptavidin-HRP
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA Development Kit |
| Quantity | 5 Plates |
| Species | Human |
| Conjugate | Biotin |
| For Use With (Application) | ELISA |
| Product Line | R&D Systems |
| Synonym | CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Research Area | Chemokines |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
