Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCL10/IP-10/CRG-2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC266RU100UG
This item is not returnable.
View return policy
Description
CXCL10/IP-10/CRG-2 Monoclonal specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CXCL10/IP-10/CRG-2 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 | |
E. coli-derived recombinant human CXCL10/IP-10/CRG-2, Val22-Pro98, Accession # P02778.2 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
33036R | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P02778.2 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
3627 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction