Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCL10/IP-10/CRG-2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC266U100UG
This item is not returnable.
View return policy
Description
CXCL10/IP-10/CRG-2 Monoclonal specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CXCL10/IP-10/CRG-2 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P02778.2 | |
CXCL10 | |
E. coli-derived recombinant human CXCL10/IP-10/CRG-2 Val22-Pro98 Accession # P02778.2 | |
RUO | |
3627 | |
Human | |
IgG1 |
Flow Cytometry | |
33036 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 | |
Mouse | |
100 μg | |
Primary | |
Detects human CXCL10/IP-10/CRG-2 in ELISAs and Western blots. In Western blots, does not cross-react with recombinant human CXCL1, 2, 3, 5, 8, 9, 12/SDF-1 beta , recombinant mouse CXCL1, or 2. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction