Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL10/IP-10/CRG-2 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC266N100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC266N100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC266N100UG Supplier R&D Systems Supplier No. IC266N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL10/IP-10/CRG-2 Monoclonal specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CXCL10/IP-10/CRG-2
Applications Flow Cytometry
Classification Monoclonal
Clone 33036
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P02778.2
Gene Alias C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
Gene Symbols CXCL10
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL10/IP-10/CRG-2 Val22-Pro98 Accession # P02778.2
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3627
Test Specificity Detects human CXCL10/IP-10/CRG-2 in ELISAs and Western blots. In Western blots, does not cross-react with recombinant human CXCL1, 2, 3, 5, 8, 9, 12/SDF-1 beta , recombinant mouse CXCL1, or 2.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.