Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCL10/IP-10/CRG-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$227.50 - $2110.00
Specifications
| Antigen | CXCL10/IP-10/CRG-2 |
|---|---|
| Clone | 33036 |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.5-2.0 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, Flow Cytometry, CyTOF, ELISA |
| Classification | Monoclonal |
Description
CXCL10/IP-10/CRG-2 Monoclonal specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
| CXCL10/IP-10/CRG-2 | |
| Western Blot 1 ug/mL, Neutralization 0.5-2.0 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P02778.2 | |
| 3627 | |
| E. coli-derived recombinant human CXCL10/IP-10/CRG-2 Val22-Pro98 Accession # P02778.2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CXCL10/IP-10/CRG-2 in ELISAs and Western blots. In Western blots, does not cross-react with recombinant human CXCL1, 2, 3, 5, 8, 9, 12/SDF-1 beta , recombinant mouse CXCL1, or 2. |
| 33036 | |
| Western Blot, Neutralization, Flow Cytometry, CyTOF, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 | |
| CXCL10 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title