Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL10/IP-10/CRG-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2661SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2661SP 25 μg
MAB2661100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2661SP Supplier R&D Systems Supplier No. MAB2661SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL10/IP-10/CRG-2 Monoclonal specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for ELISA.

Specifications

Antigen CXCL10/IP-10/CRG-2
Applications ELISA
Classification Monoclonal
Clone 33008R
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. P02778.2
Gene Alias C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL10/IP-10/CRG-2 Val22-Pro98 Accession # P02778.2
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 3627
Test Specificity Detects human CXCL10/IP-10/CRG-2 in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.