Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCL10/IP-10/CRG-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | CXCL10/IP-10/CRG-2 |
---|---|
Clone | 984429 |
Dilution | Immunohistochemistry 5-25 μg;/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2662SP
![]() |
R&D Systems
MAB2662SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2662100
![]() |
R&D Systems
MAB2662100 |
100 μg |
Each for $467.50
|
|
|||||
Description
CXCL10/IP-10/CRG-2 Monoclonal specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for Immunohistochemistry.Specifications
CXCL10/IP-10/CRG-2 | |
Immunohistochemistry 5-25 μg;/mL | |
Monoclonal | |
Supernatant | |
Human | |
P02778.2 | |
3627.0 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
984429 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 | |
E. coli-derived recombinant human CXCL10/IP-10/CRG-2, Val22-Pro98, Accession No. P02778.2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CXCL10/IP-10/CRG-2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title