Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL10/IP-10/CRG-2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF266
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF266 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF266 Supplier R&D Systems Supplier No. BAF266
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 15 publications

CXCL10/IP-10/CRG-2 Polyclonal specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen CXCL10/IP-10/CRG-2
Applications Flow Cytometry, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Alias C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
Gene Symbols CXCL10
Host Species Goat
Immunogen E. coli-derived recombinant human CXCL10/IP-10
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3627
Test Specificity Detects human CXCL10/IP-10/CRG-2 in ELISAs and Western blots. In sandwich ELISAs, less than 0.1% cross-reactivity with recombinant mouse CRG-2, recombinant human (rh) Lymphotactin, rhGRO alpha , and rhMIG is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.