Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CXCL10 (IP-10) Recombinant Protein

Catalog No. PIRP8647
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP8647 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP8647 Supplier Invitrogen™ Supplier No. RP8647

Recombinant Protein

IP10 (human) Recombinant Protein, RP-8647, from Invitrogen.

The chemokine IP-10 (or CXCL10) is a chemokine made by monocytes, endothelial cells and fibroblasts in response to treatment with IFNgamma. IP-10 functions as a chemoattractant to cells (activated T cells and NK cells) expressing the G protein-coupled receptor, CXCR3. IP-10 plays an important role in Th1 type inflammatory diseases and autoimmune diseases such as, Hashimoto′s thyroiditis, Graves′ disease and Type 1 diabetes mellitus. Recombinant human IP-10 is a non-glycosylated protein, containing 77 amino acids, with a molecular weight of 8.5 kDa. Purity is typically greater than 97% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the ability to chemoattract human T lymphocytes at 5-40ng/mL. The specific activity of this protein is 2x10^5 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: VPLSRTVRCT CISISNQPVN PRSLEKLEII PASQFCPRVE IIATMKKKGE KRCLNPESKA IKNLLKAVSK EMSKRSP.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02778
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 0.1%TFA with no preservative
Gene ID (Entrez) 3627
Molecular Weight (g/mol) ∽8.5
Name Human CXCL10
Quantity 25 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias H-IP-10
Common Name Human CXCL10
Gene Symbol CXCL10
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitro research only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.