Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL11/I-TAC Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB672100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB672100 100 μg
MAB672SP 25 μg
MAB672500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB672100 Supplier R&D Systems Supplier No. MAB672100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 8 publications

CXCL11/I-TAC Monoclonal specifically detects CXCL11/I-TAC in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CXCL11/I-TAC
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 87328
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.2-0.8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O14625
Gene Alias beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TAC, I-TACMGC102770, SCYB9B, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11
Gene Symbols CXCL11
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL11/I-TAC Phe22-Phe94 Accession # O14625
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6373
Test Specificity Detects human CXCL11/I-TAC in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant human (rh) CXCL5, 7, 8, 9, or 10. In Western blots, this antibody shows less than 1% cross-reactivity with rhCXCL1, 2, 3, 5, 6, 7, IL-8/CXCL8, 9, 10, 13, CXCL12/SDF-1 alpha , CXCL12/SDF-1 beta , rmCXCL1, 2, 6, 9, 10, CXCL12/SDF-1 alpha , rmCXCL13, rrCXCL1, CXCL3/CINC-2 alpha , CXCL3/CINC-2 beta , or rpIL-8/CXCL8.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.