Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human CXCL12 (SDF-1a) Recombinant Protein
Click to view available options
Quantity:
500 μg
Description
CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Accession Number | P48061 |
| Concentration | 200 μg/mL |
| For Use With (Application) | Control, ELISA, Western Blot |
| Formulation | 20 mM tris with 0.5 M NaCl and no preservative; pH 8 |
| Gene ID (Entrez) | 6387 |
| Molecular Weight (g/mol) | 8 kD |
| Name | Human CXCL12 alpha (SDF-1 alpha) (aa 22-89) |
| Quantity | 500 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction