Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL12/SDF-1 beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF351NA
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF351NA 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF351NA Supplier R&D Systems Supplier No. AF351NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CXCL12/SDF-1 beta Polyclonal specifically detects CXCL12/SDF-1 beta in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CXCL12/SDF-1 beta
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 10-30 ug/mL
Gene Accession No. P48061
Gene Alias hSDF-1beta, PBSF, SDF 1beta, SDF1, SDF-1, SDF1 beta, SDF1b
Gene Symbols CXCL12
Host Species Goat
Immunogen E. coli-derived recombinant human CXCL12/SDF-1 beta Lys22-Met93 Accession # P48061
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6387
Test Specificity Detects human CXCL12/SDF-1 beta in direct ELISAs and Western blots. Neutralizes 60-80% of the biological activity of CXCL12/SDF-1 beta and does not neutralize the biological activity of SDF-1 alpha . In Western blots, less than 5% cross-reactivity with recombinant human SDF-1 alpha is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.