Learn More
Human CXCL16 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Supplier:  R&D Systems FAB976N100UG
Catalog No. FAB976N100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CXCL16 Monoclonal specifically detects CXCL16 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CXCL16
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
NP_071342
CXCL16
E. coli-derived recombinant human CXCL16 Asn49-Pro137 Accession # NP_071342
RUO
58191
Human
IgG2a
Flow Cytometry
256213
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCL16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16
Rat
100 μg
Primary
Detects human CXCL16 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha ,12/SDF-1 beta , 13, recombinant mouse CXCL1, 2, 6, 9, 10, 12/SDF-1 alpha , 13, 16, recombinant rat CXCL1, 2, 3/CINC-2 alpha , 3/CINC-2 beta , recombinant canine CXCL8, or recombinant porcine CXCL8 is observed.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only (RUO)