Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCL16 APC-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB976A
This item is not returnable.
View return policy
Description
CXCL16 Monoclonal specifically detects CXCL16 in Human samples. It is validated for Flow Cytometry.Specifications
| CXCL16 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCL16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 58191 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 256213 | |
| Flow Cytometry 10 uL/10^6 cells | |
| NP_071342 | |
| CXCL16 | |
| E. coli-derived recombinant human CXCL16 Asn49-Pro137 Accession # NP_071342 | |
| 100 Tests | |
| Primary | |
| Detects human CXCL16 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha ,12/SDF-1 beta , 13, recombinant mouse CXCL1, 2, 6, 9, 10, 12/SDF-1 alpha , 13, 16, recombinant rat CXCL1, 2, 3/CINC-2 alpha , 3/CINC-2 beta , recombinant canine CXCL8, or recombinant porcine CXCL8 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction