Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL16 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB976P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB976P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB976P Supplier R&D Systems Supplier No. FAB976P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CXCL16 Monoclonal specifically detects CXCL16 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CXCL16
Applications Flow Cytometry
Classification Monoclonal
Clone 256213
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. NP_071342
Gene Alias chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCL16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16
Gene Symbols CXCL16
Host Species Rat
Immunogen E. coli-derived recombinant human CXCL16 Asn49-Pro137 Accession # NP_071342
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58191
Test Specificity Detects human CXCL16 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha ,12/SDF-1 beta , 13, recombinant mouse CXCL1, 2, 6, 9, 10, 12/SDF-1 alpha , 13, 16, recombinant rat CXCL1, 2, 3/CINC-2 alpha , 3/CINC-2 beta , recombinant canine CXCL8, or recombinant porcine CXCL8 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.