Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCL17/VCC-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | CXCL17/VCC-1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
CXCL17/VCC-1 Polyclonal specifically detects CXCL17/VCC-1 in Human samples. It is validated for Western Blot.Specifications
CXCL17/VCC-1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
Chemokine (C-X-C Motif) Ligand 17, C-X-C Motif Chemokine 17, CXCL17, Dcip1, Dendritic Cell And Monocyte Chemokine-Like Protein, DMC, UNQ473, VCC1, VCC-1, VEGF Coregulated Chemokine 1, VEGF Co-Regulated Chemokine 1 | |
CXCL17 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q6UXB2 | |
284340 | |
E. coli-derived recombinant human DMC Leu24-Leu119 Accession # Q6UXB2 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human DMC in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse DMC is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title