Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL4/PF4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB795SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB795SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB795SP Supplier R&D Systems Supplier No. MAB795SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL4/PF4 Monoclonal specifically detects CXCL4/PF4 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen CXCL4/PF4
Applications Western Blot
Classification Monoclonal
Clone 170106
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 10 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P02776.2
Gene Alias chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF4, PF-4, platelet factor 4, SCYB4oncostatin-A
Gene Symbols PF4
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL4/PF4 Glu32-Ser101 Accession # P02776.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5196
Test Specificity Detects human CXCL4/PF4 in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse CXCL4/PF4, recombinant human (rh) GRO alpha , or rhGRO beta .
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.