Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL4/PF4 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 6 publications

$200.50 - $2020.00

Specifications

Antigen CXCL4/PF4
Clone 170131
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB7951SP
GREENER_CHOICE
View Documents
R&D Systems
MAB7951SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB7951100
GREENER_CHOICE
View Documents
R&D Systems
MAB7951100
100 μg
Each for $573.00
Only null left
Add to Cart
 
MAB7951500
GREENER_CHOICE
View Documents
R&D Systems
MAB7951500
500 μg
Each for $2,020.00
Only null left
Add to Cart
 
Description

Description

CXCL4/PF4 Monoclonal specifically detects CXCL4/PF4 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

CXCL4/PF4
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF4, PF-4, platelet factor 4, SCYB4oncostatin-A
PF4
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
170131
ELISA
Unconjugated
Mouse
Human
P02776.2
5196
E. coli-derived recombinant human CXCL4/PF4 Glu32-Ser101 Accession # P02776.2
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human CXCL4/PF4 in ELISAs. In ELISAs, this antibody does not cross-react with recombinant human (rh) CXCL1, rhCXCL2, or rmCXCL4.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.