Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL4/PF4 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC7952P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC7952P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC7952P Supplier R&D Systems Supplier No. IC7952P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL4/PF4 Monoclonal specifically detects CXCL4/PF4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CXCL4/PF4
Applications Flow Cytometry
Classification Monoclonal
Clone 170138
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P02776.2
Gene Alias chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF4, PF-4, platelet factor 4, SCYB4oncostatin-A
Gene Symbols PF4
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL4/PF4 Glu32-Ser101 Accession # P02776.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5196
Test Specificity Detects human CXCL4/PF4 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) CXCL6, recombinant human (rh) CXCL6, rhCXCL7, rmCXCL13 or rhCCL21 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.