Learn More
Description
Specifications
Specifications
| Antigen | CXCL5/ENA-78 |
| Applications | CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Development, Flow Cytometry (Intracellular Staining), Mass Cytometry, Neutralization |
| Classification | Monoclonal |
| Clone | 33170 |
| Conjugate | Unconjugated |
| Dilution | Neutralization 0.65 to 5 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2 to 8 μg/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P42830 |
| Gene Alias | chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, CXCL5, ENA78, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78 |
| Gene Symbols | CXCL5 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

