Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL5/ENA-78 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 5 publications

$188.28 - $2098.77

Specifications

Antigen CXCL5/ENA-78
Clone 33160
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 3-15 ug/mL
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB254SP
GREENER_CHOICE
View Documents
R&D Systems
MAB254SP
25 μg
Each for $188.28
Only null left
Add to Cart
 
MAB254100
GREENER_CHOICE
View Documents
R&D Systems
MAB254100
100 μg
Each for $591.44
Only null left
Add to Cart
 
MAB254500
GREENER_CHOICE
View Documents
R&D Systems
MAB254500
500 μg
Each for $2,098.77
Only null left
Add to Cart
 
Description

Description

CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Specifications

Specifications

CXCL5/ENA-78
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 3-15 ug/mL
Monoclonal
Mouse
Human
P42830
6374
E. coli-derived recombinant human CXCL5/ENA-78 Ala37-Asn114 (predicted) Accession # P42830
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human CXCL5/ENA-78 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CXCL1, 2, 3, 8, 10, or recombinant mouse CXCL2 is observed.
33160
Western Blot, Immunohistochemistry, Neutralization
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, CXCL5, ENA78, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78
CXCL5
IgG1
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.