Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCL5/ENA-78 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | CXCL5/ENA-78 |
---|---|
Clone | 33103 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Human samples. It is validated for Western Blot.Specifications
CXCL5/ENA-78 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, CXCL5, ENA78, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78 | |
CXCL5 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
33103 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P42830 | |
6374 | |
E. coli-derived recombinant human CXCL5/ENA-78 Ala37-Asn114 Accession # P42830 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CXCL5/ENA-78 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CXCL1, 2, 3, 4, 6, 7, 8, 9, 10, 11, 12, 13, 16, recombinant mouse CXCL1, 2, 4, 6, 7, 9, 10, 11, 12, 15, 16, recombinant rat CXCL5, and recombinant porcine CXCL8 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title