Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCL6/GCP-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF333
This item is not returnable.
View return policy
Description
CXCL6/GCP-2 Polyclonal specifically detects CXCL6/GCP-2 in Human samples. It is validated for Western Blot, Neutralization.Specifications
CXCL6/GCP-2 | |
Polyclonal | |
Unconjugated | |
P80162 | |
CXCL6 | |
E. coli-derived recombinant human CXCL6/GCP-2 Val40-Asn114 Accession # P80162 | |
100 μg | |
Primary | |
Detects human CXCL6/GCP-2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) MIP-2, rmKC, recombinant rat (rr) CINC-2 alpha , rmCRG-2, recombinant human (rh) NAP-2, rrCINC-2 beta and rhMCP-3 is observed. | |
Human | |
IgG |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.8-4.0 ug/mL | |
chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2), Chemokine alpha 3, CKA-3Granulocyte chemotactic protein 2, GCP-2, GCP2member 6 (granulocytechemotactic protein 2), GCP-2Small-inducible cytokine B6, Small inducible cytokine subfamily B (Cys-X-Cys), member b | |
Goat | |
Affinity Purified | |
RUO | |
6372 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction