Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human CXCL8/IL-8 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP183743
Click to view available options
Quantity:
96 Tests

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Human CXCL8/IL-8 ELISA Kit (Colorimetric) measures human CXCL8/IL-8 in cell culture supernatant, serum, plasma (edta, heparin).

Specifications

Assay Range 15.6 - 1,000pg/mL
Assay Sensitivity 2.0pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3576
Gene Symbol IL8
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-8/NAP-1, Biotin-Conjugate anti-human IL-8/NAP-1 polyclonal antibody, Streptavidin-HRP, Human IL-8/NAP-1 Standard, 100ng/mL, Control high, lyophilized, Control low, lyophilized, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Angiogenesis, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor
Target CXCL8/IL-8
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.