Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL9/MIG Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC392U100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC392U100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC392U100UG Supplier R&D Systems Supplier No. IC392U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL9/MIG Monoclonal specifically detects CXCL9/MIG in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CXCL9/MIG
Applications Flow Cytometry
Classification Monoclonal
Clone 49106
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q07325
Gene Alias chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIG, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma
Gene Symbols CXCL9
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL9/MIG Thr23-Thr125 Accession # Q07325
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4283
Test Specificity Detects human CXCL9/MIG in ELISAs and Western blots. In ELISAs, does not cross-react with recombinant mouse (rm) CXCL9, recombinant human CXCL10.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.