Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCR1/IL-8RA Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FB330RN100U
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB330RN100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB330RN100U Supplier R&D Systems Supplier No. FAB330RN100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCR1/IL-8RA Monoclonal specifically detects CXCR1/IL-8RA in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CXCR1/IL-8RA
Applications Flow Cytometry
Classification Monoclonal
Clone 42705R
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide
Gene Accession No. AAA59159
Gene Alias CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 RA, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RA, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CXCR1/IL-8 RA, Met1-Leu350, Accession No. AAA59159
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3577
Test Specificity Detects human CXCR1/IL-8 RA transfectants but not the parental cell line.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.