Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCR1/IL-8RA Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB330SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB330SP 25 μg
MAB330100 100 μg
MAB330500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB330SP Supplier R&D Systems Supplier No. MAB330SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 38 publications

CXCR1/IL-8RA Monoclonal specifically detects CXCR1/IL-8RA in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-reported.

Specifications

Antigen CXCR1/IL-8RA
Applications Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF
Classification Monoclonal
Clone 42705
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Neutralization 0.4-2.0 ug/mL, CyTOF-reported
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAA59159
Gene Alias CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 RA, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RA, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A
Gene Symbols CXCR1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CXCR1/IL-8 RA Met1-Leu350 Accession # AAA59159
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3577
Test Specificity Detects human CXCR1/IL-8 RA transfectants but not the parental cell line. It does not cross-react with human CXCR2.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.