Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR1/IL-8RA Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
$139.50 - $374.00
Specifications
| Antigen | CXCR1/IL-8RA |
|---|---|
| Clone | 42705 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB330F025
![]() |
R&D Systems
FAB330F025 |
25 Tests |
Each for $139.50
|
|
|||||
FAB330F100
![]() |
R&D Systems
FAB330F100 |
100 Tests |
Each for $374.00
|
|
|||||
Description
CXCR1/IL-8RA Monoclonal specifically detects CXCR1/IL-8RA in Human samples. It is validated for Flow Cytometry.Specifications
| CXCR1/IL-8RA | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Human | |
| CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 RA, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RA, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A | |
| CXCR1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human CXCR1/IL-8 RA transfectants but not the parental cell line. It does not cross-react with human CXCR2. |
| 42705 | |
| Flow Cytometry | |
| Fluorescein | |
| Mouse | |
| AAA59159 | |
| 3577 | |
| NS0 mouse myeloma cell line transfected with human CXCR1/IL-8 RA Met1-Leu350 Accession # AAA59159 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title