Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCR2/IL-8RB APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB331A100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB331A100 100 Tests
FAB331A025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB331A100 Supplier R&D Systems Supplier No. FAB331A100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

CXCR2/IL-8RB Monoclonal specifically detects CXCR2/IL-8RB in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CXCR2/IL-8RB
Applications Flow Cytometry
Classification Monoclonal
Clone 48311
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. AAB25880
Gene Alias CD182, CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, CMKAR2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 RB, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, IL8RB, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B
Gene Symbols CXCR2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CXCR2/IL-8 RB Met1-Leu355 Accession # AAB25880
Purification Method Protein A or G purified from ascites
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 3579
Test Specificity Detects human CXCR2/IL-8 RB transfected NS0 cells but not the parental cell line. It does not cross-react with CXCR1/IL-8 RA.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.