Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR3 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB160RN100UG
This item is not returnable.
View return policy
Description
CXCR3 Monoclonal specifically detects CXCR3 in Human samples. It is validated for Flow Cytometry.Specifications
CXCR3 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
NS0 mouse myeloma cell line transfected with human CXCR3, Met1-Leu368, Accession No. P49682 | |
100 μg | |
2833.0 | |
Human | |
Supernatant |
Flow Cytometry | |
49801R | |
Flow Cytometry 0.25-1 μg/106 cells | |
P49682 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human CXCR3 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction