Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $561.50
Specifications
Antigen | CXCR3 |
---|---|
Clone | 49801R |
Dilution | Flow Cytometry 0.25 μg/10^6 cells, Immunohistochemistry 0.5-25 μg/mL, Neutralization 0.3-1.5 μg/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB160RSP
![]() |
R&D Systems
MAB160RSP |
25 μg |
Each for $241.00
|
|
|||||
MAB160R100
![]() |
R&D Systems
MAB160R100 |
100 μg |
Each for $561.50
|
|
|||||
Description
CXCR3 Monoclonal specifically detects CXCR3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready.Specifications
CXCR3 | |
Flow Cytometry 0.25 μg/10^6 cells, Immunohistochemistry 0.5-25 μg/mL, Neutralization 0.3-1.5 μg/mL, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Human | |
P49682 | |
2833 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
49801R | |
Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
NS0 mouse myeloma cell line transfected with human CXCR3 Met1-Leu368 Accession # P49682 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CXCR3 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title