Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR3 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 26 publications
$186.20 - $502.50
Specifications
| Antigen | CXCR3 |
|---|---|
| Clone | 49801 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB160P025
![]() |
R&D Systems
FAB160P025 |
25 Tests |
Each for $186.20
|
|
|||||
FAB160P100
![]() |
R&D Systems
FAB160P100 |
100 Tests |
Each for $502.50
|
|
|||||
Description
CXCR3 Monoclonal specifically detects CXCR3 in Human samples. It is validated for Flow Cytometry.Specifications
| CXCR3 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Human | |
| CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
| CXCR3 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Detects human CXCR3. |
| 49801 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| P49682 | |
| 2833 | |
| NS0 mouse myeloma cell line transfected with human CXCR3 Met1-Leu368 Accession # P49682 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title