Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR4 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB173S100UG
This item is not returnable.
View return policy
Description
CXCR4 Monoclonal specifically detects CXCR4 in Human samples. It is validated for Flow Cytometry.Specifications
CXCR4 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201E, D2S201ESDF-1 receptor, FB22, Fusin, HM89, HM89NPYRL, HSY3RR, LAP3, LAP-3, LCR1, LESTR, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3R, NPY3RWHIM, NPYR, NPYRL, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor, WHIMS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7852 | |
Human | |
IgG2b |
Flow Cytometry | |
44717 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CXCR4 | |
3T3 cells transfected with human CXCR4 | |
100 μg | |
Primary | |
Reacts specifically with human and non-human cells expressing human CXCR4 (fusin) as detected by flow cytometry. It will also react with cells expressing feline CXCR4 but not rat CXCR4. This antibody does not cross-react with other chemokine receptors. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction