Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR4 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 24 publications
$197.90 - $467.50
Specifications
Antigen | CXCR4 |
---|---|
Clone | 12G5 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB170P025
![]() |
R&D Systems
FAB170P025 |
25 Tests |
Each for $197.90
|
|
|||||
FAB170P100
![]() |
R&D Systems
FAB170P100 |
100 Tests |
Each for $467.50
|
|
|||||
Description
CXCR4 Monoclonal specifically detects CXCR4 in Human samples. It is validated for Flow Cytometry.Specifications
CXCR4 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
Human | |
CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201E, D2S201ESDF-1 receptor, FB22, Fusin, HM89, HM89NPYRL, HSY3RR, LAP3, LAP-3, LCR1, LESTR, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3R, NPY3RWHIM, NPYR, NPYRL, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor, WHIMS | |
CXCR4 | |
IgG2a | |
Protein A or G purified from ascites | |
Detects human CXCR4. |
12G5 | |
Flow Cytometry | |
R-PE | |
Mouse | |
P61073 | |
7852 | |
CP-MAC-infected SUP-T1 human T cell lymphoblastic lymphoma line Accession # P61073 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title