Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR5 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB190R100UG
This item is not returnable.
View return policy
Description
CXCR5 Monoclonal specifically detects CXCR5 in Human samples. It is validated for Flow Cytometry.Specifications
CXCR5 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P32302 | |
CXCR5 | |
NS0 mouse myeloma cell line transfected with human CXCR5 Met1-Phe372 Accession # P32302 | |
RUO | |
643 | |
Human | |
IgG2b |
Flow Cytometry | |
51505 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
BLR1, BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXCR5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15 | |
Mouse | |
100 μg | |
Primary | |
Stains human CXCR5 transfectants but not the parental cell lines in flow cytometry. Does not cross-react with human CXCR2, CXCR3, or CXCR4 transfectants. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction