Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCR5 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB190N100U
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB190N100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB190N100U Supplier R&D Systems Supplier No. FAB190N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCR5 Monoclonal specifically detects CXCR5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CXCR5
Applications Flow Cytometry
Classification Monoclonal
Clone 51505
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P32302
Gene Alias BLR1, BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXCR5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15
Gene Symbols CXCR5
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CXCR5, Met1-Phe372, Accession # P32302
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 643
Test Specificity Stains human CXCR5 transfectants but not the parental cell lines in flow cytometry. Does not cross-react with human CXCR2, CXCR3, or CXCR4 transfectants.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.