Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $573.00
Specifications
Antigen | CXCR5 |
---|---|
Clone | 51505R |
Dilution | Immunohistochemistry 3-25 μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB190RSP
![]() |
R&D Systems
MAB190RSP |
25 μg |
Each for $241.00
|
|
|||||
MAB190R100
![]() |
R&D Systems
MAB190R100 |
100 μg |
Each for $573.00
|
|
|||||
Description
CXCR5 Monoclonal specifically detects CXCR5 in Human samples. It is validated for Immunohistochemistry.Specifications
CXCR5 | |
Immunohistochemistry 3-25 μg/mL | |
Monoclonal | |
Supernatant | |
Human | |
P32302 | |
643 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
51505R | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
BLR1, BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXCR5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15 | |
NS0 mouse myeloma cell line transfected with human CXCR5, Met1-Phe372, Accession # P32302 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Stains human CXCR5 transfectants but not the parental cell lines in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title