Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR5 PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$162.90 - $502.50
Specifications
Antigen | CXCR5 |
---|---|
Clone | 51505 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB190C025
![]() |
R&D Systems
FAB190C025 |
25 Tests |
Each for $162.90
|
|
|||||
FAB190C100
![]() |
R&D Systems
FAB190C100 |
100 Tests |
Each for $502.50
|
|
|||||
Description
CXCR5 Monoclonal specifically detects CXCR5 in Human samples. It is validated for Flow Cytometry.Specifications
CXCR5 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
Human | |
BLR1, BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXCR5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15 | |
CXCR5 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Stains human CXCR5 transfectants but not the parental cell lines in flow cytometry. Does not cross-react with human CXCR2, CXCR3, or CXCR4 transfectants. |
51505 | |
Flow Cytometry | |
PerCP | |
Mouse | |
P32302 | |
643 | |
NS0 mouse myeloma cell line transfected with human CXCR5 Met1-Phe372 Accession # P32302 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title